Jars, with lid
40 mm, 25 mm
Codice catalogo HECH41050004
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Jars, low form, made of glass, with notch and overlapping cover.
Specifiche
- Ø est:40 mm
- Altezza:25 mm
Jars, low form, made of glass, with notch and overlapping cover.