Large microscopy slides
Frosted slides with cut edges, 76×52 mm
Codice catalogo MARI1100220
Nome del fornitore: MARIENFELD
Vai a
Product Details & Documents
Soda lime glass microscopy slides precleaned. Also available with a silky frosted marking area of 20 mm on both sides.
Zertifizierung: IVD directive 98/79/EC.
Specifiche
- Descrizione:Frosted slides with cut edges
- L×L:76×52 mm