ContactUsLinkComponent
PLEXON AVT STINGRAY CAMERA CS-MOUNT W/FIREWIRE-B AND TRIGGER CABLES, TRIPOD ADAPTER, AND MOUNTING KIT 80 FPS CAPTURE IN SINGLE AND DUAL CAMERA MODE REMOVABLE TILTER FOR RECORDING IN INFRARED MODE 1 * 1 ST
Codice catalogo MSPPCPX/CAM-AVT-ST
Nome del fornitore:  MARKET SOURCE PART PROCESS
PLEXON AVT STINGRAY CAMERA CS-MOUNT W/FIREWIRE-B AND TRIGGER CABLES, TRIPOD ADAPTER, AND MOUNTING KIT 80 FPS CAPTURE IN SINGLE AND DUAL CAMERA MODE REMOVABLE TILTER FOR RECORDING IN INFRARED MODE 1 * 1 ST
Codice catalogo MSPPCPX/CAM-AVT-ST
Nome del fornitore:  MARKET SOURCE PART PROCESS
Frequently Bought Together