Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Ampicillin trihydrate

Ampicillin trihydrate

100 g
Codice catalogo J66514.22
Salute
Irritante
Nome del fornitore:   Thermo Fisher Scientific
Ampicillin trihydrate
Codice catalogo J66514.22
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  7177-48-2
Ampicillin trihydrate
Ampicillin trihydrate

100 g

Numero CAS:  7177-48-2
Price
SFr. 340.00
Log in per disponibilità
1 * 100 g
Product Details & Documents

Ampicillin is lactam antibiotic with an amino group side chain attached to the penicillin structure.

Commonly used to select for ampicillin resistance in mutated and transformed cells

White powder

Soluble in water. Practically insoluble in ethanol

Sinonimi:  (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
  • Numero CAS:7177-48-2
  • Formula:C₁₆H₁₉N₃O₄S·3H₂O
  • Numero MDL:MFCD00072036
  • Molecular Weight:403.45 g/mol
  • Punto di ebollizione:684 °C
  • Punto di fusione:208 °C
  • Punto di infiammabilità:>110 °C (230 °F)
  • temperatura di conservazione:Refrigerata
  • EINECS:200-709-7
Specifiche

Specifiche

  • Conf:
    100 g

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.