Rat serum
Rat serum, Sterile filtered, Frozen
Codice catalogo S2150-050
Nome del fornitore: Biowest
Vai a
Product Details & Documents
Rat serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Rat serum is used as a blocking agent in immunoassays and controls.
Specifiche
- Formato del siero:Frozen
- Sterilità:Sterile filtered
- Descrizione:Rat serum
- Seq:55