Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
HCV genotype-4

HCV genotype-4

500 µG
Codice catalogo BIRBORB81561-500
Marketsource
Nome del fornitore:   Biorbyt
HCV genotype-4
Codice catalogo BIRBORB81561-500
Nome del fornitore:  Biorbyt
Marketsource
Numero CAS:  
HCV genotype-4

500 µG

Numero CAS:  
Price
Richiedi prezzo
Log in per disponibilità
1 * 500 µG
Product Details & Documents

Documents

Specifiche

Specifiche

  • Conf:
    500 µG
  • Cat. No.:
    BIRBORB81561-500
  • Condizioni di conservazione:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    HCV Genotype-4
  • Applicazioni testate:
    SDS-PAGE
  • Purezza:
    HCV Core Genotype-4 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-4 protein was purified by proprietary chromatographic technique.
  • Formulazione:
    50mM Tris-HCl, pH-8, 60mM NaCl, 10mM glutathione, 0.25% sarkosil 50% glycerol.

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.