Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
HCV genotype-4

HCV genotype-4

100 µG
N° de catalogue BIRBORB81561-100
Marketsource
Fournisseur:   Biorbyt
HCV genotype-4
N° de catalogue BIRBORB81561-100
Fournisseur:  Biorbyt
Marketsource
Numéro CAS:  
HCV genotype-4

100 µG

Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 100 µG
Product Details & Documents

Documents

Spécifications

Spécifications

  • Conf:
    100 µG
  • Cat. No.:
    BIRBORB81561-100
  • Condizioni di conservazione:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    HCV Genotype-4
  • Applicazioni testate:
    SDS-PAGE
  • Purezza:
    HCV Core Genotype-4 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-4 protein was purified by proprietary chromatographic technique.
  • Formulazione:
    50mM Tris-HCl, pH-8, 60mM NaCl, 10mM glutathione, 0.25% sarkosil 50% glycerol.

Spécifications