Overalls, PP, ST11
bianco, L
Codice catalogo PRTWST11WHRL
Nome del fornitore: Portwest
Vai a
Product Details & Documents
This disposable lightweight overall are made from a breathable non woven PP fabric, enabling the user to keep cool while wearing the garment.
- Generous size design provides exceptional comfort
- Shields against non-hazardous dirt, dust and grime
Weight: 40 g
Specifiche
- Taglia dell'abbigliamento:L
- Colore:bianco
- Seq:03
- Cat. No.:PRTWST11WHRL
- Peso:40 g/m²
- Modello:ST11
- Applicazioni per abiti:General Purpose|Food