Orders LinkContactUsLinkComponent
Overalls, PP, ST11
Overalls, PP, ST11
bianco, M
Codice catalogo PRTWST11WHRM
Nome del fornitore:   Portwest
Overalls, PP, ST11
Codice catalogo PRTWST11WHRM
Nome del fornitore:  Portwest
Overalls, PP, ST11
Overalls, PP, ST11
bianco, M
Price
SFr. 369.00
Log in per disponibilità
1 * 120 ST
Product Details & Documents
This disposable lightweight overall are made from a breathable non woven PP fabric, enabling the user to keep cool while wearing the garment.
  • Generous size design provides exceptional comfort
  • Shields against non-hazardous dirt, dust and grime
Weight: 40 g
Specifiche
  • Taglia dell'abbigliamento:
    M
  • Colore:
    bianco
  • Seq:
    02
  • Cat. No.:
    PRTWST11WHRM
  • Peso:
    40 g/m²
  • Modello:
    ST11
  • Applicazioni per abiti:
    General Purpose|Food