Split/splitless inlet seals
Seal, gold plated, ultra inert, with washer
Codice catalogo HEWL5190-6145
Nome del fornitore: AGILENT HEWLETT PACKARD
Split/splitless inlet seals
Codice catalogo HEWL5190-6145
Nome del fornitore: AGILENT HEWLETT PACKARD
Vai a
Product Details & Documents
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
- Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
- High quality, leak-free seal
Delivery information: An inlet washer is provided with each inlet seal.
Specifiche
- Descrizione:Seal, gold plated, ultra inert, with washer
- Seq:5