Tubes in polystyrene with screw cap
10 ml, 100 mm, 16 mm, on racks sterile packed, Sterile
Supplier: KIMA SAS DI CHIARIN RENZO
Jump to:
Product Details & Documents
Polystyrene, clear, with red screw cap.
- Sterile, individually or in the rack with 108 tubes in sterile packed available
- Non graduated
- Good thermal and chemical resistance
Universal tubes with round bottom for a wide range of applications.
Documents
Specifications
- Ø est:16 mm
- Confezione:on racks sterile packed
- Tipo:Sterile
- Capacità:10 ml
- Lunghezza:100 mm