BOUFFANT CAP COLOR: BLUE SIZE: 58CM DESCRIPTION: CLEANED, LATEXFREE ELASTIC MATERIAL: POLYPROPYLENE THICKNESS: 10-12 G/M PACKAGING: 100 PCS/DOUBLE POLYBAG, 1000 PCS/CARTON 1 * 1.000 pezzi
Codice catalogo GUNA130923
Nome del fornitore: VWR Collection
Frequently Bought Together