COVERALL COLOR: WHITE SIZE: XL DESCRIPTION: ZIPPER IN FRONT, WITH HOOD, LATEX-FREE ELASTIC ON WRIST/ANKLE/WAIST MATERIAL: POLYPROPYLENE/POLYETHYLENE THICKNESS: 40 G/M PACKAGING: 1 PC/BAG, 50 PCS/CARTON 1 * 50 pezzi
Codice catalogo GUNA131032_NMJ
Nome del fornitore: VWR Collection
Frequently Bought Together