Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Mask carriers

Pocket spacing: 4,8 mm, pocket flat: 1,0 mm, 25 masks, PP, blu, 142x69x64 mm
  ENTEA102-01-0603
 :   ENTEGRIS
Mask carriers
  ENTEA102-01-0603
 :  ENTEGRIS
Mask carriers
Mask carriers

Pocket spacing: 4,8 mm, pocket flat: 1,0 mm, 25 masks, PP, blu, 142x69x64 mm

 
 
   
1 * 4 ST
 

Miscellaneous mask carriers.

 
  • Per:
    25 masks
  • Colore:
    blu
  • Descrizione:
    Pocket spacing: 4,8 mm, pocket flat: 1,0 mm
  • Materiale:
    PP
  • Seq:
    05
  • L×P×A:
    142x69x64 mm