Spray gun
Replacement cap, natural
Codice catalogo ENTE600037
Nome del fornitore: ENTEGRIS
Vai a
Product Details & Documents
High purity PFA and PTFE (all wetted parts).
- Orifice: 4,8 mm
- Connection: 1/4" or 1/2" FNPT
- Pressure: max. 5516 mbar
Used in fume hoods and wet processing stations to help protect against corrosion.
Specifiche
- Descrizione:Replacement cap, natural
- Seq:07