Orders LinkContactUsLinkComponent
Disposable filters, Optimizer® DEV and DI
Disposable filters, Optimizer® DEV and DI
DEV-L disposable filter with 3/8" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×209 mm, 0,05 µm, 3600 cm²
Codice catalogo ENTECHUZ3L0P1
Nome del fornitore:   ENTEGRIS
Disposable filters, Optimizer® DEV and DI
Codice catalogo ENTECHUZ3L0P1
Nome del fornitore:  ENTEGRIS
Disposable filters, Optimizer® DEV and DI
Disposable filters, Optimizer® DEV and DI
DEV-L disposable filter with 3/8" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×209 mm, 0,05 µm, 3600 cm²
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
These point-of-use filters meet small to mid-volume photochemical filtration needs for positive developer and DI water applications. Both Optimizer® DEV filters (for developer applications) and Optimizer® DI filters (for DI water applications) require no pre-wetting, so the filters function immediately after installation. The design also limits the handling of hazardous chemicals during the installation and promotes a healthier workplace. Do not use either filter with aromatic hydrocarbons.
  • Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
  • O-ringless connections avoid potential sources of contamination
Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UHMWPE); Supports, core and sleeve: HDPE; Disposable shell: PP
Maximum operating temperature: 60 °C
Maximum forward differential pressure: 2,8 bar at 25 °C
Maximum reverse differential pressure: 2,8 bar at 25 °C
Articles may not be available in all countries. Please contact your local Avantor office.
Specifiche
  • Superficie di filtrazione:
    3600 cm²
  • Dimensioni pori:
    0,05 µm
  • Tipo:
    DEV-L disposable filter with 3/8" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×209 mm
  • Seq:
    19