Inserts for trays H 44 series
Nonantistatic rice paper inserts, bianco, Paper, Yes
ENTEH44-001B
: ENTEGRIS
Nest between H 44 trays and/or covers.
- Precut to tray size
- Inserts provide a softer surface providing protection for more sensitive components
- Inserts are not intended to limit part migration
- Inserts with antistatic coatings are not recommended for use when storing devices for extended periods
- Colore:bianco
- Descrizione:Nonantistatic rice paper inserts
- Materiale:Paper
- Seq:03
- Standard:Yes