Orders LinkContactUsLinkComponent
Inserts for trays H 44 series
Nonantistatic rice paper inserts, bianco, Paper, Yes
  ENTEH44-001B
 :   ENTEGRIS
Inserts for trays H 44 series
  ENTEH44-001B
 :  ENTEGRIS
Inserts for trays H 44 series
Nonantistatic rice paper inserts, bianco, Paper, Yes
1 * 1 SET
Nest between H 44 trays and/or covers.
  • Precut to tray size
  • Inserts provide a softer surface providing protection for more sensitive components
  • Inserts are not intended to limit part migration
  • Inserts with antistatic coatings are not recommended for use when storing devices for extended periods
  • Colore:
    bianco
  • Descrizione:
    Nonantistatic rice paper inserts
  • Materiale:
    Paper
  • Seq:
    03
  • Standard:
    Yes