Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

HPLC columns, Acclaim™ Trinity™ P2

Analitica, 3 µm, 3,00 mm, 100 mm
Catalog # 555-7343
HPLC columns, Acclaim™ Trinity™ P2
Catalog # 555-7343
HPLC columns, Acclaim™ Trinity™ P2
HPLC columns, Acclaim™ Trinity™ P2

Analitica, 3 µm, 3,00 mm, 100 mm

Price
SFr. 1’830.00
Log in for availability
1 * 1 ST
Product Details & Documents

The Acclaim™ Trinity™ P2 is a high-performance, silica-based column specifically designed for separation of charge molecules, including pharmaceutical counter ions by HPLC.

  • Desired selectivity for pharmaceutical counterion screening
  • Retention of ionic and ionisable analytes without using ion-pairing reagents
  • Compatibility with Thermo Scientific™ Dionex™ Corona™ Veo™ Charged Aerosol Detector (CAD) and MS detection methods

Particle type: ultra pure, spherical silica

Max. pressure: 6000 psi

Flow rate range: 0,30 to 0,90 ml/min for 3,0 mm ID formats; 0,15 to 0,45 ml/min for 2,1 mm ID formats

Bonded phase: anion-exchange, cation exchange and HILIC

pH range: 2,0 to 8,0

Specifications
  • Formato della colonna:
    Analitica
  • Ø int.:
    3,00 mm
  • Dimensioni particelle:
    3 µm
  • Lunghezza:
    100 mm
Accessories

Product Information

  • Catalog #
    Descrizione
    Availability
    Price
  • 554-4528
    Stainless steel guard cartridge holder, V-2
    Log in for availability
    1 * 1 ST - 576.00 SFr.