HPLC columns, Acclaim™ Trinity™ P2
Analitica, 3 µm, 3,00 mm, 50 mm
Codice catalogo 555-7344
Nome del fornitore: Thermo Fisher Scientific
HPLC columns, Acclaim™ Trinity™ P2
Codice catalogo 555-7344
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
The Acclaim™ Trinity™ P2 is a high-performance, silica-based column specifically designed for separation of charge molecules, including pharmaceutical counter ions by HPLC.
- Desired selectivity for pharmaceutical counterion screening
- Retention of ionic and ionisable analytes without using ion-pairing reagents
- Compatibility with Thermo Scientific™ Dionex™ Corona™ Veo™ Charged Aerosol Detector (CAD) and MS detection methods
Particle type: ultra pure, spherical silica
Max. pressure: 6000 psi
Flow rate range: 0,30 to 0,90 ml/min for 3,0 mm ID formats; 0,15 to 0,45 ml/min for 2,1 mm ID formats
Bonded phase: anion-exchange, cation exchange and HILIC
pH range: 2,0 to 8,0
Specifiche
- Formato della colonna:Analitica
- Ø int.:3,00 mm
- Dimensioni particelle:3 µm
- Lunghezza:50 mm
- Seq:03
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Stainless steel guard cartridge holder, V-2Stainless steel guard cartridge holder, V-2554-4528Log in per disponibilità1 * 1 ST - SFr. 576.00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



