Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
HPLC columns, Acclaim™ Trinity™ P2

HPLC columns, Acclaim™ Trinity™ P2

Analitica, 3 µm, 2,10 mm, 50 mm
Codice catalogo 555-7346
Nome del fornitore:   Thermo Fisher Scientific
HPLC columns, Acclaim™ Trinity™ P2
Codice catalogo 555-7346
Nome del fornitore:  Thermo Fisher Scientific
HPLC columns, Acclaim™ Trinity™ P2
HPLC columns, Acclaim™ Trinity™ P2

Analitica, 3 µm, 2,10 mm, 50 mm

Price
1 430.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents

The Acclaim™ Trinity™ P2 is a high-performance, silica-based column specifically designed for separation of charge molecules, including pharmaceutical counter ions by HPLC.

  • Desired selectivity for pharmaceutical counterion screening
  • Retention of ionic and ionisable analytes without using ion-pairing reagents
  • Compatibility with Thermo Scientific™ Dionex™ Corona™ Veo™ Charged Aerosol Detector (CAD) and MS detection methods

Particle type: ultra pure, spherical silica

Max. pressure: 6000 psi

Flow rate range: 0,30 to 0,90 ml/min for 3,0 mm ID formats; 0,15 to 0,45 ml/min for 2,1 mm ID formats

Bonded phase: anion-exchange, cation exchange and HILIC

pH range: 2,0 to 8,0

Specifiche
  • Formato della colonna:
    Analitica
  • Ø int.:
    2,10 mm
  • Dimensioni particelle:
    3 µm
  • Lunghezza:
    50 mm
  • Seq:
    01
Accessories

Informazioni sul prodotto

  • Codice catalogo
    Disponibilità
    Price
  • Stainless steel guard cartridge holder, V-2
    Stainless steel guard cartridge holder, V-2
    554-4528
    Log in per disponibilità
    1 * 1 ST - 576.00 SFr.