HPLC columns, Acclaim™ Trinity™ P2
Precartuccia, 3 µm, 2,10 mm, 10 mm
Codice catalogo 555-7342
Nome del fornitore: Thermo Fisher Scientific
HPLC columns, Acclaim™ Trinity™ P2
Codice catalogo 555-7342
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
The Acclaim™ Trinity™ P2 is a high-performance, silica-based column specifically designed for separation of charge molecules, including pharmaceutical counter ions by HPLC.
- Desired selectivity for pharmaceutical counterion screening
- Retention of ionic and ionisable analytes without using ion-pairing reagents
- Compatibility with Thermo Scientific™ Dionex™ Corona™ Veo™ Charged Aerosol Detector (CAD) and MS detection methods
Particle type: ultra pure, spherical silica
Max. pressure: 6000 psi
Flow rate range: 0,30 to 0,90 ml/min for 3,0 mm ID formats; 0,15 to 0,45 ml/min for 2,1 mm ID formats
Bonded phase: anion-exchange, cation exchange and HILIC
pH range: 2,0 to 8,0
Specifiche
- Formato della colonna:Precartuccia
- Ø int.:2,10 mm
- Dimensioni particelle:3 µm
- Lunghezza:10 mm
- Seq:05
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Stainless steel guard cartridge holder, V-2Stainless steel guard cartridge holder, V-2554-4528Log in per disponibilità1 * 1 ST - SFr. 576.00



