Mobile storage cabinets
Mobile storage cabinet with 1 shelf, 1 door, left hinged, 600×516×740 mm
Codice catalogo KOTT315-00067
Nome del fornitore: KOETTERMANN
Vai a
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifiche
- Descrizione:Mobile storage cabinet with 1 shelf
- Seq:11
- L×P×A:600×516×740 mm
- Porte:1 door, left hinged