Mobile storage cabinets
Mobile storage cabinet, with 1 drawer of 150 mm height, lockable, 1 door, right hinged, 600×516×740 mm
Codice catalogo KOTT315-00074
Nome del fornitore: KOETTERMANN
Vai a
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifiche
- Descrizione:Mobile storage cabinet, with 1 drawer of 150 mm height, lockable
- Seq:18
- L×P×A:600×516×740 mm
- Porte:1 door, right hinged