Mobile storage cabinets
Mobile storage cabinet, with 3 drawers of 150 mm height and 1 pencil tray, lockable, 3 drawers, 600×516×590 mm
Codice catalogo KOTT315-00064
Nome del fornitore: KOETTERMANN
Vai a
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifiche
- Descrizione:Mobile storage cabinet, with 3 drawers of 150 mm height and 1 pencil tray, lockable
- Seq:08
- L×P×A:600×516×590 mm
- Porte:3 drawers