Mobile storage cabinets
Mobile storage cabinet, with 3 drawers of 150 mm height, lockable, 3 drawers, 600×516×590 mm
Catalog # KOTT315-00063
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Descrizione:Mobile storage cabinet, with 3 drawers of 150 mm height, lockable
- Seq:07
- L×P×A:600×516×590 mm
- Porte:3 drawers