Mobile storage cabinets
Mobile storage cabinet, 1 door, right hinged, 600×516×590 mm
Catalog # KOTT315-00061
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Descrizione:Mobile storage cabinet
- Seq:05
- L×P×A:600×516×590 mm
- Porte:1 door, right hinged