(-)-Indolactam V ≥97%, solid
1 mg
Codice catalogo CAYM14647-1
Nome del fornitore: Cayman Chemical
(-)-Indolactam V ≥97%, solid
Codice catalogo CAYM14647-1
Nome del fornitore: Cayman Chemical
Numero CAS: 90365-57-4
Vai a
Product Details & Documents
(−)-Indolactam V is a tumor-promoting teleocidin compound that activates protein kinase C (PKC). It binds to PKC regulatory domains of mouse skin PKCη and rat brain PKCγ with Ki values of 3,4 nM and 1 μM, respectively. At 1 μM, (−)-indolactam V induces differentiation of human embryonic stem cells into pancreatic progenitors through activation of PKC signaling.
- Numero CAS:90365-57-4
- Formula:C₁₇H₂₃N₃O₂
- Molecular Weight:301.39 g/mol
Specifiche
Risultati dei test sulle specifiche
- Formal name:(2S,5S)-1,2,4,5,6,8-hexahydro-5-(hydroxymethyl)-1-methyl-2-(1-methylethyl)-3H-pyrrolo[4,3,2-gh]-1,4-benzodiazonin-3-one
- CAS number:90365-57-4
- Molecular formula:C17H23N3O2
- Formula weight:301,4
- Purity:≥97%
- Formulation:A white to off-white solid
- λmax:203, 229 and 301 nm
- Storage:4 °C
- Stability:≥ 2 years
Specifiche
- Conf:1 mg



