Winter jacket, Acode® Summit Winter
grigio scuro, 3XL
Codice catalogo KWIN1101489413XL
Vai a
Discontinued
Product Details & Documents
Wind- and waterproof polyester jackets with soft outer fabric and taped seams. Ideal for outdoor use on colder days. The jackets for women have a tailored fit.
- Discrete reflective contrasting details
- Red mesh inner lining, detachable hood
- Laminated YKK zips and zip pullers in black rubber
- Two front pockets with zips, inner pocket with zip and exit for mobile earpiece, large inner mesh pocket, sleeve pocket with zip
- Ventilation openings at armpits with zips
- Detachable snow stop inside
- Adjustable drawstring at hem, adjustable cuff straps
Waterproof rating 10000 mm
Breathability 5000 mm
Documents
Specifiche
- Taglia dell'abbigliamento:3XL
- Colore:grigio scuro
- Seq:18
- Tessuto:Polyester
- Cat. No.:KWIN1101489413XL
- Genere:Per l'uomo
- Sleeve or leg length:Long
- Risk area:Cold
- Modello:Summit Winter
- Applicazioni per abiti:General Purpose
- Tipo di chiusura:Con chiusura a zip