Winter jacket, Acode® Summit Winter
blu scuro, L
Codice catalogo KWIN110148543L
Nome del fornitore: FRISTADS KANSAS
Winter jacket, Acode® Summit Winter
Codice catalogo KWIN110148543L
Nome del fornitore: FRISTADS KANSAS
Vai a
Product Details & Documents
Wind- and waterproof polyester jackets with soft outer fabric and taped seams. Ideal for outdoor use on colder days. The jackets for women have a tailored fit.
- Discrete reflective contrasting details
- Red mesh inner lining, detachable hood
- Laminated YKK zips and zip pullers in black rubber
- Two front pockets with zips, inner pocket with zip and exit for mobile earpiece, large inner mesh pocket, sleeve pocket with zip
- Ventilation openings at armpits with zips
- Detachable snow stop inside
- Adjustable drawstring at hem, adjustable cuff straps
Waterproof rating 10000 mm
Breathability 5000 mm
Documents
Specifiche
- Taglia dell'abbigliamento:L
- Colore:blu scuro
- Seq:03
- Tessuto:Polyester
- Cat. No.:KWIN110148543L
- Genere:Per l'uomo
- Sleeve or leg length:Long
- Risk area:Cold
- Modello:Summit Winter
- Applicazioni per abiti:General Purpose
- Tipo di chiusura:Con chiusura a zip
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.