Combi top racks
30/20/12/10 mm, verde fluorescente, 4/12/32/32, 50/15/1,5/0,5 ml, 180×90×52 mm
Catalog # KOEHA-1182
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Jump to:
Product Details & Documents
Racks, made of polypropylene, neon coloured racks have four sides that can be used individually. with temperature resistant from −80 to 121 °C.
- Temperature resistant from −80 to 121 °C.
- Excellent chemical resistance
- Sturdy
Specifications
- Per provette:50/15/1,5/0,5 ml
- Ø foro:30/20/12/10 mm
- N. di fori:4/12/32/32
- Colore:verde fluorescente
- Seq:02
- L×P×A:180×90×52 mm