n-Eptano
Glass bottle, 2,5 L
1.04360.2500
: Merck
n-Heptane for gas chromatography ECD and FID SupraSolv®
: high purity solvents
- :142-82-5
- :H₃C(CH₂)₅CH₃
- :MFCD00009544
- :100.20 g/mol
- :13,04679
- :98,4 °C (1013 hPa)
- :–90,6 °C
- :0,6838 g/cm³ (20 °C)
- :–4 °C
- :Ambiente
- :1206
- :3,II
- :M-Clarity™ quality level = MQ100
- :205-563-8
- Tipo di conf.:Glass bottle
- Conf:2,5 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



