Cleaning system for controlled environment
CE Dip-N-Go sieve, grey, in stainless steel, 535×172×52 mm
Codice catalogo VILE141928
Nome del fornitore: FHP VILEDA SCS
Cleaning system for controlled environment
Codice catalogo VILE141928
Nome del fornitore: FHP VILEDA SCS
Vai a
Product Details & Documents
These cleanroom cleaning trolleys are specially designed for use in sensitive areas and controlled environments. The cleaning trolleys are extremely functional, versatile and are characterised by superior ergonomic properties.
- Small and compact
- Heat and chemical resistant
- With electrically conductive castors, the tread of the wheels is made from an hydrolysis resistant material
- Fully autoclavable and suitable for gamma sterilisation
ISO 5 / GMP A + B / D FED209 class 100
: ISO 5 / GMP A + B / D FED209 class 100
Specifiche
- Descrizione:CE Dip-N-Go sieve, grey, in stainless steel
- Dimensioni:535×172×52 mm
- Seq:11