Full length storage cabinets
Full length cabinet on leveller legs, glass door and 4 shelves, 1, 500×600×2100 mm
Codice catalogo PALBJG0554
Nome del fornitore: Palbam
Vai a
Product Details & Documents
Fabrication materials: stainless steel 304 with electropolished finish
- Removable shelves and sloping roof design as standard
- Available on either casters or leveler legs
- Assembly is quick and easy requiring no special tools or expertise
Confezione: Typically available from stock for direct shipment from the supplier. Designed to be shipped flat-packed for self-assembly. Assembly instructions are included with all cabinets.
Documents
Palbam Class - Catalog 2018
Letteratura sul prodotto
Specifiche
- Descrizione:Full length cabinet on leveller legs, glass door and 4 shelves
- Seq:01
- L×P×A:500×600×2100 mm
- Porte:1