ContactUsLinkComponent
THE PALL™ VECTOR SYSTEM WAS DEVELOPED SO THAT FOLLOWING DIGESTION WITH NSI1 AN RNA CAN BE GENERATED USING SP6 RNA POLYMERASE. 1 * 10 µG
Catalog # BIOO5205-07
Supplier:  BIOO SCIENTIFIC
THE PALL™ VECTOR SYSTEM WAS DEVELOPED SO THAT FOLLOWING DIGESTION WITH NSI1 AN RNA CAN BE GENERATED USING SP6 RNA POLYMERASE. 1 * 10 µG
Catalog # BIOO5205-07
Supplier:  BIOO SCIENTIFIC
Frequently Bought Together