THE D-POP™ KIT IS USED TO CAPTURE DNA-CONTAINING PARTICLES SUCH AS APOPTOTIC BODIES FROM CELL-FREE BIOLOGICAL SAMPLES INCLUDING BLOO D SERUM AND PLASMA, URINE, AND EUKARYOTIC CELL CULTURE MEDIA, AND TO EXTRACT DNA FROM THE CAPTURED PARTICLES. 1 * 1 KIT
BIOO5147-01
Frequently Bought Together