Shield™ DNA/RNA Reagent
DNA/RNA Shield™ (1X Concentrate), 50 ml
Codice catalogo ZYMOR1100-50
Marketsource
Nome del fornitore: Zymo Research
Shield™ DNA/RNA Reagent
Codice catalogo ZYMOR1100-50
Nome del fornitore: Zymo Research
Marketsource
Numero CAS:
Vai a
Product Details & Documents
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures.
- Nucleic acid preservation (at ambient temperature; cold-free)
- Pathogen inactivation (bacteria, fungus, parasites & viruses)
- RNA Stability: Ambient temperature (4 to 25 °C) >1 month
- DNA Stability: Ambient temperature (4 to 25 °C) >2 years
- DNA and RNA: Frozen (<–20 °C): Indefinitely
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures. There is no need for refrigeration or specialised equipment. DNA/RNA Shield™ effectively lyses cells and inactivates nucleases and infectious agents (virus), and it is compatible with various collection and storage devices (vacutainers, swabs, nasal, buccal, fecal, etc.).
Specifiche
Specifiche
- Descrizione:DNA/RNA Shield™ (1X Concentrate)
- Volume:50 ml
- Cat. No.:ZYMOR1100-50
- Prep size:Scalable
- Refine target molecule:Total RNA|Total DNA
- Target molecule:Copurification of DNA & RNA|DNA|RNA
- Method / Format:Chemical extraction & preservation
- Sample type:Stool|Swabs|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
- Descrizione:DNA/RNA Shield™ (1X Concentrate)
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



