Orders LinkContactUsLinkComponent
96-Well Blocks
96-Well Blocks
96-well block, 10 blocks, 2 ml
Codice catalogo ZYMOP1001-10
Marketsource
Nome del fornitore:   Zymo Research
96-Well Blocks
Codice catalogo ZYMOP1001-10
Nome del fornitore:  Zymo Research
Marketsource
96-Well Blocks
96-Well Blocks
96-well block, 10 blocks, 2 ml
Price
SFr. 97.00
Log in per disponibilità
1 * 10 ST
Product Details & Documents
Deep-well block, 2 ml for processing and mixing of samples.
  • Conforms to laboratory standards
96-well block features durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.
Specifiche
  • Confezione:
    10 blocks
  • Descrizione:
    96-well block
  • Seq:
    03
  • Volume totale:
    2 ml