96-Well Blocks
96-well block, 10 blocks, 2 ml
Codice catalogo ZYMOP1001-10
Marketsource
Nome del fornitore: Zymo Research
Vai a
Product Details & Documents
Deep-well block, 2 ml for processing and mixing of samples.
- Conforms to laboratory standards
96-well block features durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.
Specifiche
- Confezione:10 blocks
- Descrizione:96-well block
- Seq:03
- Volume totale:2 ml