96-Well Blocks
96-well block with cover foils, 2 blocks/foils, 2 ml
Codice catalogo ZYMOP1002-2
Marketsource
Nome del fornitore: Zymo Research
Vai a
Product Details & Documents
Deep-well block, 2 ml for processing and mixing of samples.
- Conforms to laboratory standards
96-well block features durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.
Specifiche
- Confezione:2 blocks/foils
- Descrizione:96-well block with cover foils
- Seq:02
- Volume totale:2 ml