Quick-DNA™ Kits
Product Details & Documents
DNA from cells, swabs, and whole blood.
- Quick & easy: Simple 20 minute procedure
- Unique technology excludes the use of proteinase K and organic denaturants
- Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more
The Quick-DNA™ kits are ideal for easy, rapid isolation of total DNA (e.g., genomic, mitochondrial, viral) from a variety of biological sample sources. Whole blood (fresh or stored), buffy coat, buccal cells, cells from culture, saliva, and other biological liquid samples can be processed with these kits. Zymo-Spin™ column/plate technology enables high-quality DNA purification in minutes. PCR inhibitors are effectively removed, and the eluted DNA is ideal for PCR, nucleotide blotting, DNA sequencing, restriction endonuclease digestion, bisulfite conversion/methylation analysis, and other downstream applications.
Specifiche
Specifiche
- Conf:1 KIT
- Descrizione:Quick-DNA™ 96 kit, 4×96 preps
- Cat. No.:ZYMOD3011
- Prep size:Mini
- Refine target molecule:Total DNA
- Target molecule:DNA
- Method / Format:Multiwell plate
- Sample type:Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
- Resa:Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
- Descrizione:Quick-DNA™ 96 kit
- Capacità campioni:5 µg
- Dimensione del campione:≤1×10⁶ cells or 5 mg tissue
Specifiche
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



