Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Proteinase K

Proteinase K

1×5 mg, 1 SET
Codice catalogo ZYMOD3001-2-5
Marketsource
Nome del fornitore:   Zymo Research
Proteinase K
Codice catalogo ZYMOD3001-2-5
Nome del fornitore:  Zymo Research
Marketsource
Numero CAS:  
Proteinase K
Proteinase K

1×5 mg, 1 SET

Numero CAS:  
Price
SFr. 31.00
Log in per disponibilità
1 * 1 SET
Product Details & Documents

Proteinase K is a stable serine protease with broad substrate specificity and will degrade many proteins in their native conformation, even in the presence of detergents (e.g., SDS).

  • Specifications: Lyophilized enzyme provided with Proteinase K Storage Buffer.
  • Enzyme Commission Number: (EC 3.4.21.64)
  • Source: Engyodontium album pH and Temperature Range: 4,0 to 12,0 (8,0 is optimum), 25 to 65 °C.
  • Specific Activity: Over 30 units/mg protein
  • Unit Definition: One unit (U) of enzyme will hydrolyze urea-denatured hemoglobin to produce 1.0 μmole of tyrosine per minute at pH 7,5 at 37 °C.

The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.

Specifiche

Specifiche

  • Conf:
    1 SET
  • Pk Info:
    1×5 mg
  • Cat. No.:
    ZYMOD3001-2-5
  • Nome dell'enzima:
    Proteinase K

Specifiche