Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Zymo-Spin™ IIC Columns

Zymo-Spin™ IIC Columns

Zymo-Spin™ IIC, Spin Column, 50
Catalog # ZYMOC1011-50
Marketsource
Supplier:   Zymo Research
Zymo-Spin™ IIC Columns
Catalog # ZYMOC1011-50
Supplier:  Zymo Research
Marketsource
CAS Number:  
Zymo-Spin™ IIC Columns
Zymo-Spin™ IIC Columns

Zymo-Spin™ IIC, Spin Column, 50

CAS Number:  
Price
SFr. 80.00
Log in for availability
1 * 50 ST
Product Details & Documents

Spin columns for the purification of DNA or RNA.

  • Format: DNA/RNA binding
  • DNA binding capacity: 25 µg
  • RNA binding capacity: 50 µg
  • Compatibility: microcentrifuge, vacuum manifold

The Zymo-Spin™ IIC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IIC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 μg of DNA, and 50 μg of RNA, in ≥25 μl eluate. Capacity is 900 μl.

Specifications

Specifications

  • Conf:
    50
  • Descrizione:
    Zymo-Spin™ IIC
  • Formato:
    Spin Column

Specifications