Zymo-Spin™ IC Columns
Zymo-Spin™ IC, Spin Column, 50 pack, 50
Codice catalogo ZYMOC1004-50
Marketsource
Nome del fornitore: Zymo Research
Zymo-Spin™ IC Columns
Codice catalogo ZYMOC1004-50
Nome del fornitore: Zymo Research
Marketsource
Numero CAS:
Vai a
Product Details & Documents
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 5 µg
- RNA binding capacity: 10 µg
- Compatibility: microcentrifuge, vacuum manifold
Capped version of the Zymo-Spin™ I column. The Zymo-Spin™ IC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10 μg of RNA, in ≥ 6 μl eluate. Capacity is 800 μl.
Specifiche
Specifiche
- Tipo di conf.:50 pack
- Conf:50
- Descrizione:Zymo-Spin™ IC
- Formato:Spin Column