Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Quick-DNA™ Fecal/Soil Microbe Kits

Quick-DNA™ Fecal/soil microbe 96 kit, 2× 96 preps, 96 well spin column plate, 2 KIT
Codice catalogo ZYMOD6011
Marketsource
Nome del fornitore:   Zymo Research
Quick-DNA™ Fecal/Soil Microbe Kits
Codice catalogo ZYMOD6011
Nome del fornitore:  Zymo Research
Marketsource
Numero CAS:  
Quick-DNA™ Fecal/Soil Microbe Kits
Quick-DNA™ Fecal/Soil Microbe Kits

Quick-DNA™ Fecal/soil microbe 96 kit, 2× 96 preps, 96 well spin column plate, 2 KIT

Numero CAS:  
Price
SFr. 1’017.00
Log in per disponibilità
1 * 2 KIT
Product Details & Documents

Rapid methods for the isolation of inhibitor-free, PCR quality DNA from fecal, soil, and microbial samples in minutes including tough-to-lyse bacteria, fungi, algae, and protozoa.

  • Ultra-high density BashingBeads™ are fracture resistant and chemically inert
  • Zymo-Spin™ Column and unique filtration technologies effectively removes PCR inhibitors from the DNA product

The Quick-DNA™ Fecal/Soil Microbe Kits are designed for the simple and rapid isolation of inhibitor-free, PCR-quality DNA from microbes in soil and feces. This kit can be used to isolate DNA from variety of soils (clay, sandy, silty, peaty, chalky, and loamy) and feces (humans, birds, rats, mice, cattle and more). Tough-to-lyse bacteria, fungi, protozoa, and algae are all rapidly and efficiently lysed by bead beating with our state-of-the-art, ultra-high density BashingBeads™. Zymo-Spin™ Column or Plate technology is then used to isolate the DNA, which is subsequently filtered to remove humic acids/polyphenols that can inhibit PCR. The procedures can be performed in minutes, and there is no need for organic denaturants or proteinases. Eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, methylation detection and more.

Specifiche

Specifiche

  • Conf:
    2 KIT
  • Descrizione:
    Quick-DNA™ Fecal/soil microbe 96 kit, 2× 96 preps
  • Formato:
    96 well spin column plate
  • Cat. No.:
    ZYMOD6011
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Multiwell plate
  • Sample type:
    Stool|Soil
  • Descrizione:
    Quick-DNA™ Fecal/soil microbe 96 kit, 2× 96 preps

Specifiche