Quick-RNA™ Kits
Product Details & Documents
Isolation of RNA from cells and soft tissues.
- DNA-free: Genomic DNA removal column and DNase I included
- NGS-ready: RNA is ready for all downstream applications including next-gen sequencing, RT-qPCR, hybridisation and more
The quick-RNA kits are fast RNA isolation kits designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell and tissue samples. The procedure combines a unique buffer system with zymo-spin column technology to yield high quality total RNA (including small RNAs 17 to 200 nt) in about 10 minutes.
The procedure for these RNA isolation kits is simple and fast. Add the provided RNA lysis buffer to a sample, and then purify the RNA using the zymo-spin columns. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridisation, sequencing and more.
In addition, these RNA isolation kits can be used for the enrichment of small and large RNAs into separate fractions.
Lieferinformationen: This RNA isolation kit also includes DNase I.
Technische Daten
Technische Daten
- Conf:1 KIT
- Descrizione:Quick-RNA™ 96 kit, 4×96 preps
- Cat. No.:ZYMOR1053
- Prep size:Mini
- Refine target molecule:Total RNA
- Target molecule:RNA
- Method / Format:Multiwell plate
- Sample type:Cultured cells|Human/animal cells and tissues
- Resa:10 µg RNA (binding capacity)<br />≥ 25 µl (elution volume)
- Descrizione:Quick-RNA™ 96 Kit
- Capacità campioni:10 µg
- Dimensione del campione:≤1×10⁶ Cells or 5 mg Tissue
Technische Daten
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



