Laser protection spectacles
Blue laser protection, 200/540 nm, nero, red
Codice catalogo DIDAPOD020510
Nome del fornitore: DIDALAB
Vai a
Product Details & Documents
Single lens safety glasses protect against red and blue laser.
- For wavelengths 600/760 nm or 200/540 nm
- Goggles are adjustable, one size fits all
Specifiche
- Tipo:Blue laser protection, 200/540 nm
- Colore:nero
- Materiale lente:red