Polypropylene bottles, Corning®
Wide mouth, Polypropylene, 250 ml, GL 45
Nome del fornitore: Corning
Vai a
Product Details & Documents
Bottles are manufactured from low density polyethylene and are offered with a narrow or wide mouth design.
- Hollow glass stopper style
- Reusable
- Non sterile
The cap and dispensing tube are manufactured from polypropylene to optimize backflow of the solution. The design permits minimal turbulence of the liquid in the bottle.
Documents
Certificates
Cerca per numero di lottoSpecifiche
- Filettatura:GL 45
- Capacità:250 ml
- Descrizione:Wide mouth, Polypropylene
- Seq:04