Cleanroom tapes, PCX™
Cleanroom tape, verde scuro, -, 50,8 mm
MICRPCX-2DG
These cleanroom tapes are made of LDPE with acrylic adhesive.
- Labeling
- Sealing, wafer box sealing
- Over labeling (protect paper labels in water baths)
A low density polyethylene tape, low in halogens and leachable chlorides. Processed and packaged in a cleanroom. The polyethylene film and acrylic adhesive lessen the chance of outgassing. PCX offers excellent chemical resistance and good conformability. Works well with polycarbonates, glass and acrylics. Suitable for the following:
Length: 32,9 m
Different colours available.
- Sterili:-
- Larghezza:50,8 mm
- Colore:verde scuro
- Descrizione:Cleanroom tape
- Seq:15



