Orders LinkContactUsLinkComponent
Retractable safety scalpels
Retractable safety scalpels
No. 20, Safety scalpel, sterile, individually packed, box of 25
Codice catalogo SWAN3906
Nome del fornitore:   SWANN MORTON
Retractable safety scalpels
Codice catalogo SWAN3906
Nome del fornitore:  SWANN MORTON
Retractable safety scalpels
Retractable safety scalpels
No. 20, Safety scalpel, sterile, individually packed, box of 25
Price
SFr. 184.00
Log in per disponibilità
1 * 25 ST
Product Details & Documents
Stainless steel blade, plastic handle. These disposable safety scalpels support and enhance safe passing procedures and protocols already practiced within the operating room and theatres.
  • Colour coded for ease of blade shape recognition
  • Scalpel features a strong temporary forward lock position to prevent uncontrolled retraction when contacting with bone during use
  • Additional notch to hold the blade more securely in the passing position and a rear permanent lock to be engaged prior to final disposal in a sharps container
Confezione: All scalpels are available in single peel sterile packs or in a non sterile bulk format.
Specifiche
  • Tipo:
    No. 20
  • Descrizione:
    Safety scalpel, sterile, individually packed, box of 25
  • Seq:
    04