Retractable safety scalpels
No. 11P, Safety scalpel, sterile, individually packed, box of 25
Nome del fornitore: SWANN MORTON
Vai a
Product Details & Documents
Stainless steel blade, plastic handle. These disposable safety scalpels support and enhance safe passing procedures and protocols already practiced within the operating room and theatres.
- Colour coded for ease of blade shape recognition
- Scalpel features a strong temporary forward lock position to prevent uncontrolled retraction when contacting with bone during use
- Additional notch to hold the blade more securely in the passing position and a rear permanent lock to be engaged prior to final disposal in a sharps container
Confezione: All scalpels are available in single peel sterile packs or in a non sterile bulk format.
Specifiche
- Tipo:No. 11P
- Descrizione:Safety scalpel, sterile, individually packed, box of 25
- Seq:03