Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Multi-element standard solutions for ion chromatography

Multi-element standard solutions for ion chromatography

Multi-element standards for IC (7 components), 1000 mg/l, 100 mL
Codice catalogo ACSD1521.1K.W.L1
Nome del fornitore:   CPACHEM
Multi-element standard solutions for ion chromatography
Codice catalogo ACSD1521.1K.W.L1
Nome del fornitore:  CPACHEM
Numero CAS:  
Multi-element standard solutions for ion chromatography
Multi-element standard solutions for ion chromatography

Multi-element standards for IC (7 components), 1000 mg/l, 100 mL

Numero CAS:  
Price
245.00 SFr.
Log in per disponibilità
1 * 100 mL
Product Details & Documents

This standard solution is intended for ion chromatography.

Multi element standards for Br⁻, Cl-, F⁻, NO₂⁻, NO₃⁻, PO₄³⁻, SO₄²⁻ diluted in H₂O.

Specifiche

Specifiche

  • Concentrazione:
    1000 mg/l
  • Conf:
    100 mL
  • Descrizione:
    Multi-element standards for IC (7 components)

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.